siRNA Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289947.1 | NP_001276876.1 | 4 | 350 | ||
| NM_001290030.1 | NP_001276959.1 | 9 | 1137 | |||
| XM_011510998.2 | XP_011509300.1 | 11 | 1269 | |||
| XM_011510999.2 | XP_011509301.1 | 9 | 840 | |||
| XM_011511001.2 | XP_011509303.1 | 9 | 1615 | |||
| XM_017003875.1 | XP_016859364.1 | 10 | 1638 | |||
| XM_017003876.1 | XP_016859365.1 | 9 | 653 | |||
| GenBank mRNA | BC127676.1 | - | 4 | 350 | 40127368 | |
| BC150546.1 | - | 4 | 350 | |||
| BQ429225.1 | - | 348 | ||||

ERICH2
Genomic Map