siRNA Details
Gene Symbol: CROCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005450.1 | XP_016860939.1 | 4 | 593 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CROCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005450.1 | XP_016860939.1 | 4 | 593 |