siRNA Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242617.1 | NP_001229546.1 | 4 | 2019 | ||
| NM_001242618.1 | NP_001229547.1 | 4 | 1980 | |||
| NM_018698.4 | NP_061168.2 | 5 | 2109 | |||
| XM_011530989.2 | XP_011529291.1 | 4 | 2022 | |||
| XM_017029648.1 | XP_016885137.1 | 4 | 1957 | |||
| GenBank mRNA | AK023289.1 | - | 5 | 2109 | ||
| BC014888.1 | - | 4 | 1973 | |||

NXT2
Genomic Map