siRNA Details
Gene Symbol: LINC01097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034054.1 | 4 | 1147 | |||
GenBank mRNA | AK096368.1 | - | 4 | 1147 | ||
BC035722.1 | - | 4 | 972 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034054.1 | 4 | 1147 | |||
GenBank mRNA | AK096368.1 | - | 4 | 1147 | ||
BC035722.1 | - | 4 | 972 |