siRNA Details
| Gene Symbol: LINC01019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033898.1 | 5 | 2178 | |||
| GenBank mRNA | BC035019.1 | - | 5 | 2130 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01019| Gene Symbol: LINC01019 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033898.1 | 5 | 2178 | |||
| GenBank mRNA | BC035019.1 | - | 5 | 2130 | ||