siRNA Details
| Gene Symbol: TMEM55A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018710.2 | NP_061180.1 | 3 | 511 | ||
| XM_011517152.2 | XP_011515454.1 | 3 | 511 | |||
| GenBank mRNA | AK313783.1 | - | 3 | 424 | ||
| AX775961.1 | - | 3 | 361 | |||
| BC033892.1 | - | 3 | 361 | 4820065 | ||
| CR749733.1 | - | 2 | 75 | |||
| DA171968.1 | - | 3 | 511 | |||

TMEM55A
Genomic Map