siRNA Details
| Gene Symbol: LINC01622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027115.3 | 1, 2 | 1628 | |||
| NR_027116.2 | 1, 2 | 1628 | ||||
| GenBank mRNA | BC037855.2 | - | 1, 2 | 1629 | ||
| BC117448.1 | - | 1, 2 | 1291 | |||
| BC126314.1 | - | 1, 2 | 1291 | |||

LINC01622
Genomic Map