siRNA Details
| Gene Symbol: LOC286149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093190.1 | - | 2 | 584 | ||
| Gene Symbol: LOC105375650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132986.1 | 2 | 634 | |||
| NR_132987.1 | 2 | 584 | ||||

LOC286149
Genomic Map