siRNA Details
| Gene Symbol: H2BFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164416.1 | NP_001157888.1 | 11 | |||
| XM_006724640.2 | XP_006724703.1 | 1 | 44 | |||
| XM_011530922.2 | XP_011529224.1 | 1 | 207 | |||
| GenBank mRNA | AK093522.1 | - | 11 | |||

H2BFM
Genomic Map