siRNA Details
| Gene Symbol: H2BFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164416.1 | NP_001157888.1 | 3 | 634 | ||
| GenBank mRNA | AK093522.1 | - | 3 | 634 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2BFM| Gene Symbol: H2BFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164416.1 | NP_001157888.1 | 3 | 634 | ||
| GenBank mRNA | AK093522.1 | - | 3 | 634 | ||