siRNA Details
| Gene Symbol: TTC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310135.1 | NP_001297064.1 | 5, 6 | 2169 | ||
| XM_011537432.2 | XP_011535734.1 | 5, 6 | 2169 | |||
| XM_017021254.1 | XP_016876743.1 | 5, 6 | 2169 | |||
| XM_017021255.1 | XP_016876744.1 | 5, 6 | 2169 | |||
| XM_017021256.1 | XP_016876745.1 | 3, 4 | 2259 | |||
| XM_017021257.1 | XP_016876746.1 | 5, 6 | 2169 | |||

TTC6
Genomic Map