siRNA Details
| Gene Symbol: TMEM202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080462.1 | NP_001073931.1 | 3 | 470 | ||
| XM_011521497.1 | XP_011519799.1 | 3 | 471 | |||
| XM_011521499.1 | XP_011519801.1 | 3 | 291 | |||
| GenBank mRNA | AK131033.1 | - | 2 | 240 | ||

TMEM202
Genomic Map