siRNA Details
| Gene Symbol: PFKFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271805.1 | NP_001258734.1 | 2 | 166 | ||
| NM_002625.3 | NP_002616.2 | 3 | 331 | |||
| XM_017029576.1 | XP_016885065.1 | 3 | 735 | |||
| XM_017029577.1 | XP_016885066.1 | 3 | 735 | |||
| XM_017029578.1 | XP_016885067.1 | 4 | 817 | |||
| GenBank mRNA | AK300724.1 | - | 2 | 166 | ||
| AK314089.1 | - | 3 | 547 | |||
| BC096078.2 | - | 3 | 333 | 40000711 | ||
| BC096079.3 | - | 3 | 332 | 40000713 | ||
| X52638.1 | - | 3 | 339 | |||

PFKFB1
Genomic Map