siRNA Details
Gene Symbol: C17orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243552.1 | NP_001230481.1 | 4 | 628 | ||
XM_017024004.1 | XP_016879493.1 | 4 | 565 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C17orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243552.1 | NP_001230481.1 | 4 | 628 | ||
XM_017024004.1 | XP_016879493.1 | 4 | 565 |