siRNA Details
| Gene Symbol: C20orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199534.1 | NP_001186463.1 | 2 | 425 | ||
| NM_018840.4 | NP_061328.1 | 2 | 425 | |||
| NM_199483.2 | NP_955777.1 | 2 | 425 | |||
| NR_026562.3 | 2 | 425 | ||||
| GenBank mRNA | AF112213.1 | - | 2 | 389 | ||
| AF274936.1 | - | 2 | 320 | |||
| AF274948.1 | - | 2 | 238 | |||
| BC001871.1 | - | 2 | 342 | 3546888 | ||
| BC004446.1 | - | 2 | 350 | 2819807 | ||
| BQ086248.1 | - | 2 | 374 | |||
| BU174619.1 | - | 393 | ||||
| BU189639.1 | - | 2 | 337 | |||
| BU194101.1 | - | 352 | ||||
| BU848918.1 | - | 2 | 384 | |||
| S83364.1 | - | 2 | 165 | |||
| Gene Symbol: TGIF2-C20orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199535.1 | NP_001186464.1 | 3 | 401 | ||

C20orf24
Genomic Map