siRNA Details
| Gene Symbol: THEG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205273.2 | NP_001192202.1 | 2 | 587 | ||
| NM_001278575.1 | NP_001265504.1 | 3 | 997 | |||
| NM_001278576.1 | NP_001265505.1 | 4 | 1121 | |||
| NM_001278577.1 | NP_001265506.1 | 4 | 1132 | |||
| NM_001278578.1 | NP_001265507.1 | 3 | 990 | |||
| XM_011526302.2 | XP_011524604.1 | 3 | 2776 | |||
| XM_011526303.2 | XP_011524605.1 | 3 | 1601 | |||
| XM_017026126.1 | XP_016881615.1 | 3 | 3112 | |||
| XM_017026127.1 | XP_016881616.1 | 3 | 2571 | |||
| XM_017026128.1 | XP_016881617.1 | 3 | 2769 | |||
| XM_017026129.1 | XP_016881618.1 | 3 | 1608 | |||
| XM_017026130.1 | XP_016881619.1 | 3 | 1333 | |||
| XM_017026131.1 | XP_016881620.1 | 4 | 939 | |||
| XM_017026132.1 | XP_016881621.1 | 3 | 1132 | |||
| XM_017026133.1 | XP_016881622.1 | 2 | 800 | |||
| GenBank mRNA | BC043286.1 | - | 4 | 1060 | ||

THEG5
Genomic Map