siRNA Details
Gene Symbol: LOC339593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038972.1 | 2 | 1126 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC339593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038972.1 | 2 | 1126 |