siRNA Details
| Gene Symbol: LINC00298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015405.1 | 3 | 417 | |||
| GenBank mRNA | BC043361.1 | - | 4 | 488 | ||
| BC103700.1 | - | 3 | 417 | |||
| BC104747.1 | - | 4 | 535 | |||
| BC105688.1 | - | 3 | 417 | |||
| BC107115.1 | - | 3 | 417 | |||

LINC00298
Genomic Map