siRNA Details
| Gene Symbol: FMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103184.3 | NP_001096654.1 | 1 | 202 | ||
| GenBank mRNA | AL833157.1 | - | 1 | 333 | ||
| BC101971.1 | - | 1 | 202 | |||
| BC103692.1 | - | 1 | 202 | |||
| BC107593.1 | - | 1 | 202 | |||

FMN1
Genomic Map