siRNA Details
| Gene Symbol: SLFN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129820.1 | NP_001123292.1 | 1 | 271 | ||
| XM_017024576.1 | XP_016880065.1 | 3 | 440 | |||
| XM_017024577.1 | XP_016880066.1 | 3 | 462 | |||
| XM_017024578.1 | XP_016880067.1 | 2 | 329 | |||
| XM_017024579.1 | XP_016880068.1 | 2 | 362 | |||
| GenBank mRNA | BC140847.1 | - | 1 | 268 | 9021715 | |
| BC157877.1 | - | 1 | 272 | 9057094 | ||
| BC157879.1 | - | 1 | 279 | 9057095 | ||

SLFN14
Genomic Map