siRNA Details
| Gene Symbol: LRRC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195545.1 | NP_001182474.1 | 2 | 294 | ||
| XM_011524158.2 | XP_011522460.1 | 4 | 480 | |||
| XM_017024003.1 | XP_016879492.1 | 4 | 459 | |||
| GenBank mRNA | DY655553.1 | - | 2 | 294 | ||

LRRC3C
Genomic Map