siRNA Details
Gene Symbol: LEUTX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143832.1 | NP_001137304.1 | 1, 2 | 77 | ||
GenBank mRNA | CR746510.1 | - | 1, 2 | 76 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LEUTX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143832.1 | NP_001137304.1 | 1, 2 | 77 | ||
GenBank mRNA | CR746510.1 | - | 1, 2 | 76 |