siRNA Details
| Gene Symbol: C19orf81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195076.1 | NP_001182005.1 | 4 | 377 | ||
| XM_011526910.2 | XP_011525212.1 | 4 | 567 | |||
| GenBank mRNA | BX281890.1 | - | 3 | 320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C19orf81| Gene Symbol: C19orf81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195076.1 | NP_001182005.1 | 4 | 377 | ||
| XM_011526910.2 | XP_011525212.1 | 4 | 567 | |||
| GenBank mRNA | BX281890.1 | - | 3 | 320 | ||