siRNA Details
| Gene Symbol: ZNF878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080404.2 | NP_001073873.2 | 4 | 1298 | ||
| XM_017027195.1 | XP_016882684.1 | 3 | 1701 | |||
| XM_017027196.1 | XP_016882685.1 | 3 | 1698 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF878| Gene Symbol: ZNF878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080404.2 | NP_001073873.2 | 4 | 1298 | ||
| XM_017027195.1 | XP_016882684.1 | 3 | 1701 | |||
| XM_017027196.1 | XP_016882685.1 | 3 | 1698 | |||