siRNA Details
| Gene Symbol: ZNF878 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080404.2 | NP_001073873.2 | 4 | 411 | ||
| XM_017027195.1 | XP_016882684.1 | 3 | 814 | |||
| XM_017027196.1 | XP_016882685.1 | 3 | 811 | |||
| GenBank mRNA | BQ222464.1 | - | 4 | 383 | ||

ZNF878
Genomic Map