siRNA Details
| Gene Symbol: RGPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017004845.1 | XP_016860334.1 | 1, 2 | 101 | ||
| XM_017004848.1 | XP_016860337.1 | 1, 2 | 101 | |||
| XM_017004849.1 | XP_016860338.1 | 1, 2 | 101 | |||
| XM_017004850.1 | XP_016860339.1 | 1, 2 | 101 | |||

RGPD2
Genomic Map