siRNA Details
| Gene Symbol: DUBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028301.1 | 2 | 442 | |||
| GenBank mRNA | AK055934.1 | - | 2 | 310 | ||
| BC039550.1 | - | 2 | 410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUBR| Gene Symbol: DUBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028301.1 | 2 | 442 | |||
| GenBank mRNA | AK055934.1 | - | 2 | 310 | ||
| BC039550.1 | - | 2 | 410 | |||