siRNA Details
| Gene Symbol: IRGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145805.1 | NP_001139277.1 | 2 | 1558 | ||
| XM_011537641.2 | XP_011535943.1 | 2 | 1558 | |||
| GenBank mRNA | BC038539.1 | - | 1 | 72 | ||
| BC128167.1 | - | 1 | 438 | 40113046 | ||
| BC128168.1 | - | 1 | 445 | 40113047 | ||
| EU742619.1 | - | 2 | 1558 | |||

IRGM
Genomic Map