siRNA Details
| Gene Symbol: PCDHGC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018928.2 | NP_061751.1 | 1 | 1501 | ||
| NM_032406.1 | NP_115782.1 | 1 | 1501 | |||
| GenBank mRNA | AF152338.1 | - | 1 | 1501 | ||
| AF152525.1 | - | 1 | 1501 | |||
| BC101035.1 | - | 1 | 1501 | 40007872 | ||
| BC101036.1 | - | 1 | 1498 | |||
| BC101038.2 | - | 1 | 1501 | 40007875 | ||

PCDHGC4
Genomic Map