siRNA Details
| Gene Symbol: LINC00264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026793.1 | 1, 2 | 468 | |||
| GenBank mRNA | BC038537.1 | - | 1, 2 | 468 | ||
| Gene Symbol: LINC00202-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026794.1 | 3, 4 | 5471 | |||
| GenBank mRNA | CR749234.1 | - | 3, 4 | 5411 | ||

LINC00264
Genomic Map