siRNA Details
Gene Symbol: LINC00202-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026794.1 | 5 | 6050 | |||
GenBank mRNA | BX105108.1 | - | 2 | 520 | ||
CR749234.1 | - | 5 | 5990 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00202-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026794.1 | 5 | 6050 | |||
GenBank mRNA | BX105108.1 | - | 2 | 520 | ||
CR749234.1 | - | 5 | 5990 |