siRNA Details
| Gene Symbol: TEX36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318133.1 | NP_001305062.1 | 4 | 630 | ||
| GenBank mRNA | BX336391.2 | - | 4 | 592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX36| Gene Symbol: TEX36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318133.1 | NP_001305062.1 | 4 | 630 | ||
| GenBank mRNA | BX336391.2 | - | 4 | 592 | ||