siRNA Details
| Gene Symbol: TEX36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128202.2 | NP_001121674.1 | 3 | 396 | ||
| NM_001318133.1 | NP_001305062.1 | 3 | 396 | |||
| XM_005269817.4 | XP_005269874.1 | 3 | 394 | |||
| GenBank mRNA | AK131010.1 | - | 2 | 69 | ||
| BC062717.1 | - | 3 | 399 | 6101709 | ||
| BC127766.1 | - | 3 | 277 | 40127344 | ||
| BC127767.1 | - | 3 | 281 | 40127345 | ||
| BX336391.2 | - | 3 | 358 | |||

TEX36
Genomic Map