siRNA Details
| Gene Symbol: DNHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144666.2 | NP_653267.2 | 13 | 2935 | ||
| GenBank mRNA | BG676072.1 | - | 3 | 332 | ||
| BX647806.1 | - | 2 | 169 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNHD1| Gene Symbol: DNHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144666.2 | NP_653267.2 | 13 | 2935 | ||
| GenBank mRNA | BG676072.1 | - | 3 | 332 | ||
| BX647806.1 | - | 2 | 169 | |||