siRNA Details
| Gene Symbol: PCDHB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018936.3 | NP_061759.1 | 1 | 392 | ||
| GenBank mRNA | AB209745.1 | - | 1 | 168 | ||
| AF152495.1 | - | 1 | 208 | |||
| AF217756.1 | - | 1 | 208 | |||
| AK027526.1 | - | 1 | 364 | |||
| AK314642.1 | - | 1 | 346 | |||
| BC098347.1 | - | 1 | 255 | 40007951 | ||
| BC098575.1 | - | 1 | 356 | 30921306 | ||
| BF206341.1 | - | 1 | 345 | |||
| DA703023.1 | - | 1 | 392 | |||

PCDHB2
Genomic Map