siRNA Details
Gene Symbol: MIPEPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038939.1 | 1 | 126 | |||
NR_046461.1 | 1 | 126 | ||||
GenBank mRNA | AK093279.1 | - | 1 | 78 | ||
DA538853.1 | - | 1 | 127 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MIPEPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038939.1 | 1 | 126 | |||
NR_046461.1 | 1 | 126 | ||||
GenBank mRNA | AK093279.1 | - | 1 | 78 | ||
DA538853.1 | - | 1 | 127 |