siRNA Details
| Gene Symbol: LINC00539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA446493.1 | - | 1, 2 | 41 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00539| Gene Symbol: LINC00539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA446493.1 | - | 1, 2 | 41 | ||