siRNA Details
Gene Symbol: LINC01550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015430.1 | 3 | 1095 | |||
GenBank mRNA | BC043585.1 | - | 3 | 1095 | ||
JX073283.1 | - | 3 | 1031 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015430.1 | 3 | 1095 | |||
GenBank mRNA | BC043585.1 | - | 3 | 1095 | ||
JX073283.1 | - | 3 | 1031 |