siRNA Details
| Gene Symbol: IGHV1OR15-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135694.1 | 2 | 302 | |||
| GenBank mRNA | HY100667.1 | - | 2 | 305 | ||
| Gene Symbol: LOC102724760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135695.1 | 2 | 302 | |||
| GenBank mRNA | DN998389.1 | - | 2 | 282 | ||

IGHV1OR15-1
Genomic Map