siRNA Details
| Gene Symbol: SH3BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018957.3 | NP_061830.3 | 7 | 602 | ||
| GenBank mRNA | AK124370.1 | - | 8 | 1429 | ||
| AK126556.1 | - | 6 | 1297 | |||
| AK301163.1 | - | 7 | 584 | |||
| AY364241.1 | - | 4 | 299 | |||
| BC008282.1 | - | 5 | 395 | 3639782 | ||
| CR456576.1 | - | 7 | 521 | |||

SH3BP1
Genomic Map