siRNA Details
| Gene Symbol: GOLGA8J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282472.1 | NP_001269401.1 | 16 | 1527 | ||
| XM_006720651.3 | XP_006720714.1 | 16 | 1547 | |||
| Gene Symbol: GOLGA8H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282490.1 | NP_001269419.1 | 16 | 1525 | ||

GOLGA8J
Genomic Map