siRNA Details
| Gene Symbol: DAZAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018959.3 | NP_061832.2 | 3 | 367 | ||
| NM_170711.2 | NP_733829.1 | 3 | 367 | |||
| XM_005259531.3 | XP_005259588.1 | 3 | 320 | |||
| XM_005259534.3 | XP_005259591.1 | 3 | 326 | |||
| XM_005259535.3 | XP_005259592.1 | 3 | 328 | |||
| XM_005259536.3 | XP_005259593.1 | 3 | 328 | |||
| XM_011527904.2 | XP_011526206.1 | 3 | 1632 | |||
| XM_011527905.2 | XP_011526207.1 | 3 | 1640 | |||
| XM_011527906.2 | XP_011526208.1 | 3 | 1640 | |||
| XM_011527907.2 | XP_011526209.1 | 3 | 1649 | |||
| XM_011527908.2 | XP_011526210.1 | 3 | 1639 | |||
| XM_011527909.2 | XP_011526211.1 | 3 | 1639 | |||
| XM_011527910.2 | XP_011526212.1 | 3 | 719 | |||
| XM_017026581.1 | XP_016882070.1 | 3 | 362 | |||
| XM_017026582.1 | XP_016882071.1 | 3 | 1655 | |||
| GenBank mRNA | AF181719.1 | - | 3 | 270 | ||
| AK125032.1 | - | 2 | 1663 | |||
| AL360261.1 | - | 3 | 262 | |||
| BC012062.1 | - | 3 | 235 | 4549444 | ||
| BI909277.1 | - | 3 | 351 | |||
| BU557183.1 | - | 367 | ||||
| KU178593.1 | - | 3 | 162 | |||

DAZAP1
Genomic Map