siRNA Details
| Gene Symbol: AGBL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046012.1 | 3 | 505 | |||
| GenBank mRNA | CD359326.1 | - | 3 | 504 | ||
| DB459014.1 | - | 2 | 459 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGBL1-AS1| Gene Symbol: AGBL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046012.1 | 3 | 505 | |||
| GenBank mRNA | CD359326.1 | - | 3 | 504 | ||
| DB459014.1 | - | 2 | 459 | |||