siRNA Details
| Gene Symbol: LOC106660606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132988.1 | 2 | 236 | |||
| GenBank mRNA | BM766743.1 | - | 2 | 236 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106660606| Gene Symbol: LOC106660606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132988.1 | 2 | 236 | |||
| GenBank mRNA | BM766743.1 | - | 2 | 236 | ||