siRNA Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242757.1 | NP_001229686.1 | 5, 6 | 981 | ||
| XM_011522897.2 | XP_011521199.1 | 6, 7 | 1029 | |||
| XM_011522899.2 | XP_011521201.1 | 6, 7 | 966 | |||
| XM_017022962.1 | XP_016878451.1 | 3, 4 | 631 | |||
| XM_017022963.1 | XP_016878452.1 | 3, 4 | 2379 | |||
| XM_017022964.1 | XP_016878453.1 | 4, 5 | 1535 | |||
| XM_017022965.1 | XP_016878454.1 | 3, 4 | 2320 | |||
| XM_017022966.1 | XP_016878455.1 | 4, 5 | 2158 | |||

SLC22A31
Genomic Map