siRNA Details
| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145640.1 | NP_001139112.1 | 2 | 181 | ||
| NR_027149.1 | 3 | 339 | ||||
| GenBank mRNA | AL365406.1 | - | 3 | 325 | ||
| BC034005.1 | - | 2 | 185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFR2| Gene Symbol: ZFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145640.1 | NP_001139112.1 | 2 | 181 | ||
| NR_027149.1 | 3 | 339 | ||||
| GenBank mRNA | AL365406.1 | - | 3 | 325 | ||
| BC034005.1 | - | 2 | 185 | |||