siRNA Details
| Gene Symbol: C1orf132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135298.1 | 2 | 623 | |||
| NR_135299.1 | 3 | 808 | ||||
| GenBank mRNA | EU154351.1 | - | 2 | 623 | ||
| EU154352.1 | - | 3 | 808 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf132| Gene Symbol: C1orf132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135298.1 | 2 | 623 | |||
| NR_135299.1 | 3 | 808 | ||||
| GenBank mRNA | EU154351.1 | - | 2 | 623 | ||
| EU154352.1 | - | 3 | 808 | |||