siRNA Details
| Gene Symbol: LINC00028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024358.1 | 2, 3 | 403 | |||
| GenBank mRNA | BX280465.1 | - | 2, 3 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00028| Gene Symbol: LINC00028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024358.1 | 2, 3 | 403 | |||
| GenBank mRNA | BX280465.1 | - | 2, 3 | 403 | ||