siRNA Details
| Gene Symbol: LINC00649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038883.2 | 1 | 548 | |||
| NR_134559.1 | 1 | 548 | ||||
| GenBank mRNA | AK311545.1 | - | 1 | 548 | ||
| AL532366.3 | - | 1 | 33 | |||
| BX102617.1 | - | 1 | 507 | |||
| DA374118.1 | - | 1 | 458 | |||

LINC00649
Genomic Map