siRNA Details
| Gene Symbol: LINC00649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038883.2 | 1 | 502 | |||
| NR_134559.1 | 1 | 502 | ||||
| GenBank mRNA | AK311545.1 | - | 1 | 502 | ||
| BX102617.1 | - | 1 | 461 | |||
| DA374118.1 | - | 1 | 412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00649| Gene Symbol: LINC00649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038883.2 | 1 | 502 | |||
| NR_134559.1 | 1 | 502 | ||||
| GenBank mRNA | AK311545.1 | - | 1 | 502 | ||
| BX102617.1 | - | 1 | 461 | |||
| DA374118.1 | - | 1 | 412 | |||